Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 26 | 5180 | ||
XM_011523768.2 | XP_011522070.1 | 25 | 5111 | |||
GenBank mRNA | AB014564.1 | - | 22 | 4569 | ||
AK023003.1 | - | 18 | 4029 | |||
AK074252.1 | - | 11 | 2374 | |||
AK093541.1 | - | 1 | 2208 | |||
AK125717.1 | - | 26 | 5199 | |||
BC004266.1 | - | 8 | 2130 | |||
BC033614.1 | - | 8 | 1996 | |||
BC063786.1 | - | 20 | 4262 | |||
BX648864.1 | - | 4 | 2829 | |||
CR627449.1 | - | 24 | 4918 |