Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015336.2 | NP_056151.2 | 13 | 1517 | ||
XM_005268750.4 | XP_005268807.1 | 13 | 1316 | |||
XM_011538083.1 | XP_011536385.1 | 13 | 1381 | |||
XM_017019091.1 | XP_016874580.1 | 14 | 1467 | |||
XM_017019092.1 | XP_016874581.1 | 13 | 1330 | |||
XM_017019093.1 | XP_016874582.1 | 13 | 1279 | |||
XM_017019094.1 | XP_016874583.1 | 12 | 1197 | |||
XM_017019095.1 | XP_016874584.1 | 9 | 909 | |||
GenBank mRNA | AB023163.1 | - | 13 | 1461 | ||
AB024494.1 | - | 13 | 2007 | |||
AB097013.1 | - | 13 | 1337 | |||
AB097035.1 | - | 13 | 1337 | |||
AK292866.1 | - | 13 | 1333 | |||
BC050324.1 | - | 13 | 1476 | 5266097 |