Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003941.3 | NP_003932.3 | 3 | 658 | ||
GenBank mRNA | AK289795.1 | - | 3 | 571 | ||
AM295156.1 | - | 3 | 321 | |||
BC052955.1 | - | 3 | 608 | 5264663 | ||
CF593336.1 | - | 3 | 688 | |||
CR749438.1 | - | 4 | 563 | |||
D88460.1 | - | 3 | 575 |