Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015362.4 | NP_056177.3 | 9 | 1337 | ||
NM_203413.2 | NP_981958.1 | 7 | 2123 | |||
NM_203414.2 | NP_981959.1 | 8 | 1425 | |||
NM_203415.1 | NP_981960.1 | 9 | 1230 | |||
XM_011523779.2 | XP_011522081.1 | 9 | 1329 | |||
GenBank mRNA | AF087868.1 | - | 8 | 1098 | ||
AF163262.1 | - | 6 | 1842 | |||
AK225983.1 | - | 8 | 1340 | |||
AL713741.1 | - | 2 | 1467 | |||
BC002762.2 | - | 9 | 1302 | 3629369 | ||
BF982004.1 | - | 2 | 363 | |||
CA450318.1 | - | 4 | 559 |