Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 5128 | ||
NM_001145103.1 | NP_001138575.1 | 9 | 4939 | |||
NM_001145104.1 | NP_001138576.1 | 7 | 4572 | |||
NM_005902.3 | NP_005893.1 | 9 | 5363 | |||
XM_011521559.2 | XP_011519861.1 | 8 | 5393 | |||
XM_011521560.1 | XP_011519862.1 | 9 | 5305 | |||
GenBank mRNA | AF151027.1 | - | 1 | 611 | ||
AL110265.1 | - | 1 | 1166 | |||
BC000414.1 | - | 1 | 516 | |||
BC007496.1 | - | 1 | 516 | |||
CR749287.1 | - | 1 | 4123 |