Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145795.1 | NP_001139267.1 | 9 | 2850 | ||
NM_001145796.1 | NP_001139268.1 | 10 | 2950 | |||
NM_001145797.1 | NP_001139269.1 | 10 | 3003 | |||
NM_001145812.1 | NP_001139284.1 | 10 | 3049 | |||
NM_001308293.1 | NP_001295222.1 | 11 | 3106 | |||
NM_001308294.1 | NP_001295223.1 | 9 | 1885 | |||
NM_015503.2 | NP_056318.2 | 9 | 4780 | |||
XM_017023114.1 | XP_016878603.1 | 8 | 3269 | |||
XM_017023115.1 | XP_016878604.1 | 10 | 2976 | |||
XM_017023116.1 | XP_016878605.1 | 9 | 4334 | |||
XM_017023117.1 | XP_016878606.1 | 7 | 3065 | |||
XM_017023118.1 | XP_016878607.1 | 8 | 1956 | |||
XM_017023119.1 | XP_016878608.1 | 8 | 1785 | |||
XM_017023120.1 | XP_016878609.1 | 7 | 1619 | |||
GenBank mRNA | AB037720.1 | - | 9 | 5969 | ||
AF227967.1 | - | 9 | 2706 | |||
AF227968.1 | - | 10 | 2806 | |||
AF227969.1 | - | 10 | 2859 | |||
AK027488.1 | - | 9 | 2115 | |||
AK055104.1 | - | 11 | 3017 | |||
AL049924.1 | - | 9 | 2303 | |||
AL713760.1 | - | 9 | 4780 | |||
BC010704.1 | - | 9 | 2503 | 3866324 |