Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015527.3 | NP_056342.3 | 9 | 2360 | ||
XM_011545789.1 | XP_011544091.1 | 6 | 2102 | |||
XM_011545790.2 | XP_011544092.1 | 8 | 2227 | |||
GenBank mRNA | AB449895.1 | - | 9 | 1877 | ||
AF370370.1 | - | 9 | 1506 | |||
AK002101.1 | - | 3 | 863 | |||
AK023192.1 | - | 9 | 1739 | |||
AL117408.1 | - | 2 | 1557 | |||
AY007150.1 | - | 1 | 79 | |||
BC050523.2 | - | 5 | 1074 | |||
BC063112.1 | - | 9 | 1784 | |||
BC072453.1 | - | 9 | 1672 | 6184429 | ||
BC093814.1 | - | 9 | 1588 | 7939659 | ||
BC093816.1 | - | 9 | 1588 | 7939661 | ||
BC098419.1 | - | 9 | 1924 |