Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 1851 | ||
XM_011515596.2 | XP_011513898.1 | 13 | 1367 | |||
XM_017012746.1 | XP_016868235.1 | 14 | 1858 | |||
XM_017012747.1 | XP_016868236.1 | 14 | 1891 | |||
GenBank mRNA | AF308300.1 | - | 14 | 1528 | ||
AK294857.1 | - | 14 | 1800 |
Gene Symbol: PLEKHA8P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037144.1 | 3 | 1638 | |||
GenBank mRNA | AF103731.1 | - | 3 | 1638 | ||
AF380162.1 | - | 1 | 1467 | |||
AK024218.1 | - | 1 | 613 | |||
BC028914.1 | - | 1 | 1479 | |||
BC063575.1 | - | 3 | 1519 | 3881751 |