Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 29 | 4193 | ||
NM_016284.4 | NP_057368.3 | 29 | 4208 | |||
NM_206999.2 | NP_996882.1 | 29 | 4208 | |||
NR_049763.1 | 29 | 4193 | ||||
GenBank mRNA | AB023224.2 | - | 16 | 2253 | ||
AK302495.1 | - | 6 | 578 | |||
AL117492.1 | - | 1 | 47 | |||
AL832769.1 | - | 29 | 4157 | |||
AL833549.1 | - | 29 | 4167 | |||
BC024317.1 | - | 12 | 1607 | |||
BC040523.1 | - | 29 | 4160 | 5266600 | ||
BX537840.1 | - | 29 | 4139 | |||
BX647815.1 | - | 29 | 4148 | |||
CR749237.1 | - | 22 | 3148 | |||
EF553522.1 | - | 29 | 4142 |