Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164372.1 | NP_001157844.1 | 6, 7 | 859 | ||
NM_001164373.1 | NP_001157845.1 | 6, 7 | 768 | |||
NM_016301.3 | NP_057385.3 | 6, 7 | 738 | |||
XM_005253896.4 | XP_005253953.3 | 7, 8 | 1193 | |||
XM_017019394.1 | XP_016874883.1 | 6, 7 | 673 | |||
GenBank mRNA | AF117229.1 | - | 6, 7 | 688 | ||
AK223209.1 | - | 6, 7 | 710 | |||
AK314946.1 | - | 6, 7 | 711 | |||
AM992619.1 | - | 6, 7 | 657 | |||
AM992620.1 | - | 6, 7 | 657 | |||
AY359078.1 | - | 6, 7 | 739 | |||
AY364262.1 | - | 6, 7 | 657 | |||
BC008416.1 | - | 6, 7 | 713 | 4048678 | ||
BC031024.1 | - | 6, 7 | 714 | 4695834 | ||
BX537973.1 | - | 6, 7 | 749 |