Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113546.1 | NP_001107018.1 | 4 | 623 | ||
NM_001113547.1 | NP_001107019.1 | 1 | 504 | |||
NM_016357.4 | NP_057441.1 | 4 | 623 | |||
XM_011538455.2 | XP_011536757.1 | 4 | 683 | |||
XM_017019430.1 | XP_016874919.1 | 4 | 537 | |||
GenBank mRNA | AB209512.1 | - | 4 | 559 | ||
AF198454.1 | - | 4 | 550 | |||
AF198455.1 | - | 1 | 442 | |||
AK000335.1 | - | 1 | 147 | |||
AK000372.1 | - | 4 | 533 | |||
AK222971.1 | - | 4 | 546 | |||
AK297766.1 | - | 2 | 219 | |||
AK314447.1 | - | 4 | 535 | |||
AL136911.1 | - | 4 | 542 | |||
BC001247.1 | - | 1 | 155 | |||
BC110815.1 | - | 4 | 560 | 5115521 | ||
BC136763.1 | - | 4 | 509 | 9020753 | ||
BC144402.1 | - | 4 | 509 | |||
BC144403.1 | - | 4 | 509 | |||
DA738195.1 | - | 1 | 504 |
Gene Symbol: C5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130086.1 | - | 1671 |