Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113546.1 | NP_001107018.1 | 6, 7 | 1030 | ||
NM_001113547.1 | NP_001107019.1 | 3, 4 | 911 | |||
NM_016357.4 | NP_057441.1 | 6, 7 | 1030 | |||
XM_011538455.2 | XP_011536757.1 | 6, 7 | 1090 | |||
GenBank mRNA | AB209512.1 | - | 6, 7 | 966 | ||
AF198454.1 | - | 6, 7 | 957 | |||
AF198455.1 | - | 3, 4 | 849 | |||
AF218025.1 | - | 2, 3 | 164 | |||
AK000335.1 | - | 3, 4 | 554 | |||
AK000372.1 | - | 6, 7 | 940 | |||
AK222971.1 | - | 6, 7 | 953 | |||
AK297766.1 | - | 4, 5 | 626 | |||
AK314447.1 | - | 6, 7 | 942 | |||
AL136911.1 | - | 6, 7 | 949 | |||
BC001247.1 | - | 3, 4 | 562 | |||
BC110815.1 | - | 6, 7 | 967 | 5115521 | ||
BC136763.1 | - | 6, 7 | 916 | 9020753 | ||
BC144402.1 | - | 6, 7 | 916 | |||
BC144403.1 | - | 6, 7 | 916 |