Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 6 | 1247 |
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199885.1 | NP_001186814.1 | 3 | 748 | ||
NM_016400.3 | NP_057484.3 | 4 | 804 | |||
GenBank mRNA | AK022435.1 | - | 2 | 711 | ||
BC019262.1 | - | 4 | 802 | 3504222 |