Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324066.1 | NP_001310995.1 | 3, 4 | 728 | ||
NM_016641.3 | NP_057725.1 | 3, 4 | 715 | |||
NR_136689.1 | 3, 4 | 732 | ||||
GenBank mRNA | AF212862.1 | - | 3, 4 | 671 | ||
AF306698.1 | - | 3, 4 | 624 | |||
AK312912.1 | - | 3, 4 | 685 | |||
AY463154.1 | - | 3, 4 | 551 | |||
BC014981.1 | - | 3, 4 | 606 | 4855079 | ||
BC025273.1 | - | 3, 4 | 666 | 5427491 | ||
BQ897064.1 | - | 3, 4 | 715 |