Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 5, 6 | 751 | ||
NM_199139.2 | NP_954590.1 | 4, 5 | 694 | |||
NR_046398.1 | 3, 4 | 498 | ||||
XM_005256706.4 | XP_005256763.1 | 5, 6 | 1323 | |||
XM_005256708.2 | XP_005256765.1 | 6, 7 | 1313 | |||
XM_011523945.2 | XP_011522247.1 | 6, 7 | 1138 | |||
XM_011523946.2 | XP_011522248.1 | 6, 7 | 1138 | |||
XM_011523947.2 | XP_011522249.1 | 5, 6 | 659 | |||
XM_017024780.1 | XP_016880269.1 | 5, 6 | 952 | |||
GenBank mRNA | AK056908.1 | - | 5, 6 | 1547 | ||
AK290276.1 | - | 7, 8 | 821 | |||
AK292710.1 | - | 5, 6 | 531 | |||
AK292848.1 | - | 4, 5 | 474 | |||
BC032776.1 | - | 3, 4 | 281 | |||
BC058017.1 | - | 4, 5 | 461 | |||
BC073156.1 | - | 5, 6 | 550 | 6068243 | ||
BM475564.1 | - | 5, 6 | 522 | |||
BX648588.1 | - | 4, 5 | 1082 | |||
BX649188.1 | - | 5, 6 | 2998 | |||
X99699.1 | - | 5, 6 | 509 |