Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 7 | 2373 | ||
NM_199139.2 | NP_954590.1 | 6 | 2316 | |||
NR_046396.1 | 6 | 2287 | ||||
NR_046397.1 | 5 | 2230 | ||||
NR_046398.1 | 5 | 2120 | ||||
XM_005256706.4 | XP_005256763.1 | 7 | 2945 | |||
XM_005256708.2 | XP_005256765.1 | 8 | 2935 | |||
XM_011523945.2 | XP_011522247.1 | 8 | 2760 | |||
XM_011523947.2 | XP_011522249.1 | 7 | 2281 | |||
XM_017024780.1 | XP_016880269.1 | 7 | 2574 | |||
GenBank mRNA | AK091799.1 | - | 1 | 727 | ||
BX648588.1 | - | 6 | 2703 | |||
BX649188.1 | - | 7 | 4620 |