Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF821 SIRNA Assays ›
Gene Symbol:
ZNF821
Gene Name:
zinc finger protein 821
Gene Aliases:
-
Chromosome Location:
Chr.16: 71859680 - 71886224 on Build GRCh38
UniGene ID:
Hs.744055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140392
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF821
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201552.1
NP_001188481.1
5, 6
740
NM_001201553.1
NP_001188482.1
4, 5
725
NM_001201554.1
NP_001188483.1
4, 5
614
NM_001201556.1
NP_001188485.1
5, 6
740
NM_001318239.1
NP_001305168.1
4, 5
467
NM_017530.2
NP_060000.1
3, 4
599
XM_005256033.2
XP_005256090.1
6, 7
814
XM_006721233.1
XP_006721296.1
5, 6
673
XM_011523211.2
XP_011521513.1
4, 5
519
XM_017023409.1
XP_016878898.1
5, 6
658
XM_017023410.1
XP_016878899.1
5, 6
1405
XM_017023411.1
XP_016878900.1
5, 6
482
XM_017023412.1
XP_016878901.1
6, 7
814
XM_017023414.1
XP_016878903.1
4, 5
601
XM_017023416.1
XP_016878905.1
5, 6
751
XM_017023417.1
XP_016878906.1
5, 6
745
XM_017023418.1
XP_016878907.1
5, 6
607
XM_017023419.1
XP_016878908.1
5, 6
589
XM_017023420.1
XP_016878909.1
6, 7
834
XM_017023422.1
XP_016878911.1
3, 4
255
XM_017023423.1
XP_016878912.1
5, 6
748
XM_017023424.1
XP_016878913.1
4, 5
602
GenBank mRNA
AF070588.1
-
3, 4
567
AK296603.1
-
4, 5
725
AK313693.1
-
4, 5
574
AL538838.3
-
5, 6
759
BC012116.1
-
4, 5
571
4553333
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline