Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017748.4 | NP_060218.1 | 5 | 745 | ||
NR_073428.1 | 6 | 862 | ||||
GenBank mRNA | AK000298.1 | - | 5 | 695 | ||
AK225373.1 | - | 5 | 715 | |||
AK296115.1 | - | 6 | 827 | |||
AK302983.1 | - | 5 | 646 | |||
AK316077.1 | - | 4 | 499 | |||
BC008833.2 | - | 5 | 731 | 4304656 |