Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017748.4 | NP_060218.1 | 10 | 1966 | ||
NR_073428.1 | 11 | 2083 | ||||
GenBank mRNA | AK000298.1 | - | 10 | 1914 | ||
AK225373.1 | - | 10 | 1936 | |||
BC008833.2 | - | 10 | 1952 | 4304656 | ||
CR749362.1 | - | 6 | 1148 |