Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017758.3 | NP_060228.3 | 4 | 2065 | ||
GenBank mRNA | AK000129.1 | - | 2 | 513 | ||
AK000315.1 | - | 4 | 1416 | |||
AK311161.1 | - | 4 | 792 | |||
AK311511.1 | - | 4 | 750 | |||
AL137263.1 | - | 2 | 770 | |||
BC062339.1 | - | 5 | 1594 | 6339184 |