Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017758.3 | NP_060228.3 | 4 | 3287 | ||
GenBank mRNA | AK000129.1 | - | 2 | 1735 | ||
AK025817.1 | - | 1 | 1212 | |||
AL137263.1 | - | 2 | 1993 | |||
BC062339.1 | - | 5 | 2817 | 6339184 |