Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305544.1 | NP_001292473.1 | 10 | 3211 | ||
NM_001305545.1 | NP_001292474.1 | 9 | 2293 | |||
NM_017763.5 | NP_060233.3 | 10 | 3414 | |||
GenBank mRNA | AB081837.1 | - | 10 | 2951 | ||
AK000322.1 | - | 9 | 2631 | |||
AK291027.1 | - | 10 | 3184 | |||
AK296769.1 | - | 8 | 2404 | |||
BC109028.2 | - | 9 | 2660 | 40027722 | ||
CR627423.1 | - | 10 | 3211 | |||
CR749257.1 | - | 9 | 4419 |