Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319988.1 | NP_001306917.1 | 9 | 1448 | ||
NM_001319993.1 | NP_001306922.1 | 9 | 1325 | |||
NM_001319994.1 | NP_001306923.1 | 8 | 1212 | |||
NM_017915.4 | NP_060385.3 | 8 | 1217 | |||
XM_011538509.1 | XP_011536811.1 | 7 | 983 | |||
XM_011538514.2 | XP_011536816.1 | 7 | 1030 | |||
XM_011538515.1 | XP_011536817.1 | 6 | 823 | |||
XM_017019534.1 | XP_016875023.1 | 7 | 1046 | |||
XM_017019535.1 | XP_016875024.1 | 7 | 1056 | |||
XM_017019538.1 | XP_016875027.1 | 8 | 1217 | |||
XM_017019541.1 | XP_016875030.1 | 9 | 1306 | |||
GenBank mRNA | AK000648.1 | - | 8 | 1136 | ||
AK302724.1 | - | 9 | 1434 | |||
AK303571.1 | - | 8 | 1143 | |||
BC030962.1 | - | 9 | 1269 | |||
BC070270.1 | - | 8 | 1173 | |||
BC098313.1 | - | 5 | 607 | 40025466 |