Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003986.2 | NP_003977.1 | 3 | 448 | ||
XM_005253159.2 | XP_005253216.1 | 3 | 696 | |||
XM_005253160.2 | XP_005253217.1 | 3 | 693 | |||
XM_005253161.2 | XP_005253218.1 | 3 | 445 | |||
XM_011520402.1 | XP_011518704.1 | 4 | 856 | |||
XM_017018403.1 | XP_016873892.1 | 2 | 378 | |||
GenBank mRNA | AF082868.1 | - | 2 | 146 | ||
AK313422.1 | - | 2 | 196 | |||
BC011034.1 | - | 1 | 136 | 4042355 | ||
CR456812.1 | - | 1 | 80 | |||
DA268467.1 | - | 3 | 446 |