Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018011.3 | NP_060481.3 | 1, 2 | 588 | ||
GenBank mRNA | AK001016.1 | - | 1, 2 | 586 | ||
AK303575.1 | - | 2, 3 | 361 | |||
AK304751.1 | - | 2, 3 | 418 | |||
BC050434.1 | - | 1, 2 | 535 | 6016552 | ||
BC071587.1 | - | 1, 2 | 559 | 5271043 |