Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 18, 19 | 2449 | ||
GenBank mRNA | AK001167.1 | - | 8, 9 | 908 | ||
AK025479.1 | - | 12, 13 | 1482 | |||
AK056433.1 | - | 18, 19 | 2459 | |||
AK093941.1 | - | 5, 6 | 1428 | |||
AK297564.1 | - | 18, 19 | 2234 | |||
AK307766.1 | - | 17, 18 | 2299 | |||
AL137396.1 | - | 2, 3 | 240 | |||
BC000536.2 | - | 8, 9 | 913 | |||
BC007214.1 | - | 13, 14 | 1571 | |||
BC110072.1 | - | 5, 6 | 585 | |||
BC110852.1 | - | 13, 14 | 1504 | |||
BC125108.1 | - | 18, 19 | 2381 | 40118772 | ||
BC125109.1 | - | 18, 19 | 2381 | 40118777 | ||
CR457343.1 | - | 12, 13 | 1417 |