Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040616.2 | NP_001035706.1 | 2 | 533 | ||
XM_005254941.1 | XP_005254998.1 | 2 | 481 | |||
XM_005254943.1 | XP_005255000.1 | 1 | 413 | |||
XM_017022398.1 | XP_016877887.1 | 1 | 413 | |||
XM_017022400.1 | XP_016877889.1 | 2 | 737 | |||
GenBank mRNA | AB083157.1 | - | 1 | 363 | ||
AK001445.1 | - | 2 | 467 | |||
AK292972.1 | - | 2 | 464 | |||
BI910880.1 | - | 3 | 563 | |||
DA447859.1 | - | 2 | 533 | |||
DA674923.1 | - | 2 | 492 | |||
DA880742.1 | - | 2 | 477 | |||
DA958361.1 | - | 2 | 481 |