Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040616.2 | NP_001035706.1 | 4 | 725 | ||
XM_005254941.1 | XP_005254998.1 | 4 | 673 | |||
XM_005254943.1 | XP_005255000.1 | 3 | 605 | |||
XM_017022398.1 | XP_016877887.1 | 4 | 767 | |||
XM_017022399.1 | XP_016877888.1 | 2 | 142 | |||
XM_017022400.1 | XP_016877889.1 | 5 | 1091 | |||
GenBank mRNA | AB083157.1 | - | 3 | 555 | ||
AK001445.1 | - | 4 | 659 | |||
AK292972.1 | - | 4 | 656 | |||
AK298946.1 | - | 4 | 304 | |||
DA880742.1 | - | 4 | 669 |