Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077498.2 | NP_001070966.1 | 3 | 2969 | ||
NM_001288631.1 | NP_001275560.1 | 4 | 3140 | |||
NM_001288632.1 | NP_001275561.1 | 5 | 3051 | |||
NM_001288633.1 | NP_001275562.1 | 4 | 3045 | |||
NM_001288634.1 | NP_001275563.1 | 3 | 3038 | |||
NM_001288635.1 | NP_001275564.1 | 4 | 3017 | |||
NM_001288636.1 | NP_001275565.1 | 6 | 3041 | |||
NM_001288637.1 | NP_001275566.1 | 5 | 3007 | |||
NM_001288638.1 | NP_001275567.1 | 5 | 2939 | |||
NM_001288639.1 | NP_001275568.1 | 4 | 2891 | |||
NM_001288640.1 | NP_001275569.1 | 3 | 2857 | |||
NM_018182.3 | NP_060652.2 | 4 | 3003 | |||
XM_011524986.2 | XP_011523288.1 | 3 | 2934 | |||
XM_011524987.1 | XP_011523289.1 | 4 | 2769 | |||
XM_011524988.2 | XP_011523290.1 | 3 | 2985 | |||
XM_017024831.1 | XP_016880320.1 | 4 | 3093 | |||
XM_017024832.1 | XP_016880321.1 | 3 | 3358 | |||
XM_017024833.1 | XP_016880322.1 | 3 | 2898 | |||
XM_017024834.1 | XP_016880323.1 | 3 | 3096 | |||
XM_017024835.1 | XP_016880324.1 | 4 | 3295 | |||
GenBank mRNA | AK001562.1 | - | 4 | 2955 | ||
CB156081.1 | - | 1 | 230 |