Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC47A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018242.2 | NP_060712.2 | 16 | 1504 | ||
GenBank mRNA | AK001709.1 | - | 16 | 1504 | ||
AK222625.1 | - | 16 | 1508 | |||
AK225360.1 | - | 6 | 576 | |||
AK298254.1 | - | 7 | 746 | |||
AK301854.1 | - | 14 | 1366 | |||
AK304647.1 | - | 8 | 683 | |||
BC010661.2 | - | 16 | 1493 | 3874616 | ||
BC050592.1 | - | 16 | 1478 | |||
BC058882.1 | - | 13 | 1380 | 5188651 |