Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099455.1 | NP_001092925.1 | 3 | 597 | ||
NM_018340.2 | NP_060810.2 | 4 | 1023 | |||
GenBank mRNA | AK002013.1 | - | 4 | 1006 | ||
AK022710.1 | - | 4 | 973 | |||
AK093854.1 | - | 4 | 769 | |||
AK225359.1 | - | 4 | 973 | |||
AK225617.1 | - | 4 | 1018 | |||
AK298662.1 | - | 3 | 427 | |||
AY553877.1 | - | 4 | 912 | |||
BC006289.2 | - | 4 | 1007 | 3959701 | ||
BX640805.1 | - | 3 | 563 |