Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099455.1 | NP_001092925.1 | 3 | 2136 | ||
NM_018340.2 | NP_060810.2 | 4 | 2562 | |||
GenBank mRNA | AK002013.1 | - | 4 | 2545 | ||
AK093854.1 | - | 4 | 2308 | |||
BC006289.2 | - | 4 | 2546 | 3959701 |