Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018346.2 | NP_060816.1 | 9 | 1782 | ||
NR_130911.1 | 5 | 1421 | ||||
GenBank mRNA | AK002026.1 | - | 9 | 1753 | ||
AK022105.1 | - | 9 | 1722 | |||
AK024628.1 | - | 4 | 762 | |||
BC005854.2 | - | 8 | 1534 | |||
BC050538.1 | - | 9 | 1713 | 5214885 |