Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099642.1 | NP_001093112.1 | 3 | 879 | ||
NM_001324374.1 | NP_001311303.1 | 3 | 810 | |||
NM_001324377.1 | NP_001311306.1 | 3 | 581 | |||
NM_001324378.1 | NP_001311307.1 | 2 | 834 | |||
NM_001324379.1 | NP_001311308.1 | 2 | 896 | |||
NM_018348.5 | NP_060818.4 | 3 | 1023 | |||
XM_011523228.1 | XP_011521530.1 | 3 | 1168 | |||
XM_011523229.1 | XP_011521531.1 | 3 | 1163 | |||
XM_017023444.1 | XP_016878933.1 | 3 | 1366 | |||
GenBank mRNA | AF458590.1 | - | 3 | 923 | ||
AK002033.1 | - | 1 | 607 | |||
AK023183.1 | - | 1 | 312 | |||
AK226169.1 | - | 3 | 774 | |||
AK315056.1 | - | 3 | 821 | |||
BC034468.1 | - | 3 | 605 | 4824516 | ||
BC035005.2 | - | 3 | 810 | 5267637 | ||
BX648660.1 | - | 3 | 786 |