Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018405.3 | NP_060875.2 | 3 | 298 | ||
XM_005258000.1 | XP_005258057.1 | 3 | 408 | |||
GenBank mRNA | AF407672.1 | - | 3 | 190 | ||
AJ272196.2 | - | 3 | 205 | |||
AK024772.1 | - | 3 | 218 | |||
BC012386.1 | - | 3 | 228 | 3882737 | ||
BC035483.1 | - | 4 | 430 | |||
DB472863.1 | - | 3 | 328 |