Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 11 | 1031 | ||
NM_018427.4 | NP_060897.3 | 12 | 1121 | |||
XM_005255376.1 | XP_005255433.1 | 10 | 896 | |||
XM_005255377.4 | XP_005255434.1 | 9 | 845 | |||
XM_011522540.2 | XP_011520842.1 | 7 | 533 | |||
XM_011522541.1 | XP_011520843.1 | 11 | 960 | |||
GenBank mRNA | AF227156.1 | - | 12 | 1087 | ||
AJ272050.1 | - | 12 | 1041 | |||
AK225662.1 | - | 12 | 1097 | |||
AK295486.1 | - | 9 | 714 | |||
AK300253.1 | - | 11 | 940 | |||
AK302994.1 | - | 9 | 768 | |||
AK303405.1 | - | 8 | 614 | |||
AK304856.1 | - | 12 | 987 | |||
AK314769.1 | - | 12 | 1035 | |||
AK316520.1 | - | 11 | 945 | |||
BC071868.1 | - | 4 | 356 | |||
BC132688.1 | - | 12 | 1038 | 40146710 | ||
BC132690.1 | - | 12 | 1038 | 40146712 | ||
DA764092.1 | - | 4 | 324 |