Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018838.4 | NP_061326.1 | 3 | 218 | ||
GenBank mRNA | AF112208.1 | - | 3 | 238 | ||
AF217092.1 | - | 3 | 197 | |||
AK222619.1 | - | 3 | 205 | |||
BC005936.1 | - | 3 | 207 | 4069306 | ||
BG943528.1 | - | 3 | 203 | |||
BM548072.1 | - | 285 | ||||
BT007220.1 | - | 3 | 185 | |||
U34343.1 | - | 324 |