Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258338.1 | NP_001245267.1 | 3 | 257 | ||
NM_018838.4 | NP_061326.1 | 4 | 345 | |||
GenBank mRNA | AF112208.1 | - | 4 | 365 | ||
AF217092.1 | - | 4 | 324 | |||
AK222619.1 | - | 4 | 332 | |||
BG943528.1 | - | 4 | 330 | |||
BM548072.1 | - | 412 | ||||
BU532909.1 | - | 3 | 244 | |||
U34343.1 | - | 452 |