Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142640.1 | NP_001136112.1 | 4 | 1609 | ||
NM_018996.3 | NP_061869.2 | 4 | 1609 | |||
XM_006721996.3 | XP_006722059.3 | 5 | 7261 | |||
XM_006721997.3 | XP_006722060.3 | 5 | 7261 | |||
XM_006722000.3 | XP_006722063.3 | 5 | 7261 | |||
XM_011525088.2 | XP_011523390.1 | 2 | 1298 | |||
XM_017024906.1 | XP_016880395.1 | 5 | 7261 | |||
XM_017024907.1 | XP_016880396.1 | 5 | 7261 | |||
XM_017024908.1 | XP_016880397.1 | 5 | 7261 | |||
GenBank mRNA | AB046802.2 | - | 1 | 1159 |