Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020233.4 | NP_064618.3 | 4 | 1098 | ||
XM_005256738.2 | XP_005256795.1 | 4 | 1094 | |||
GenBank mRNA | AB073393.1 | - | 4 | 1069 | ||
BC001294.2 | - | 3 | 1137 | 3452502 | ||
BC070155.1 | - | 4 | 1181 | 4656998 | ||
DA280275.1 | - | 1 | 138 |