Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308347.1 | NP_001295276.1 | 6 | 807 | ||
NM_001308358.1 | NP_001295287.1 | 6 | 846 | |||
NM_020313.3 | NP_064709.2 | 6 | 846 | |||
XM_005256063.4 | XP_005256120.1 | 6 | 698 | |||
GenBank mRNA | AF116609.1 | - | 4 | 457 | ||
AF248964.1 | - | 6 | 750 | |||
AK074263.1 | - | 6 | 687 | |||
AK292281.1 | - | 6 | 650 | |||
AL136613.1 | - | 6 | 668 | |||
BC002568.2 | - | 6 | 688 | 3139226 | ||
BC024196.2 | - | 6 | 727 | 4816594 | ||
BC067303.1 | - | 6 | 846 | 30389410 | ||
BC071740.1 | - | 6 | 721 | 5563094 | ||
BG746807.1 | - | 1 | 692 | |||
CR533545.1 | - | 5 | 565 |