Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308347.1 | NP_001295276.1 | 9 | 1344 | ||
NM_001308358.1 | NP_001295287.1 | 8 | 1267 | |||
NM_020313.3 | NP_064709.2 | 9 | 1383 | |||
XM_005256063.4 | XP_005256120.1 | 8 | 1119 | |||
GenBank mRNA | AF116609.1 | - | 7 | 994 | ||
AF248964.1 | - | 9 | 1287 | |||
AK074263.1 | - | 9 | 1224 | |||
AK292281.1 | - | 9 | 1187 | |||
AL136613.1 | - | 9 | 1205 | |||
BC002568.2 | - | 9 | 1225 | 3139226 | ||
BC024196.2 | - | 9 | 1264 | 4816594 | ||
BC067303.1 | - | 9 | 1383 | 30389410 | ||
BC071740.1 | - | 9 | 1258 | 5563094 |