Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308347.1 | NP_001295276.1 | 9 | 1737 | ||
NM_001308358.1 | NP_001295287.1 | 8 | 1660 | |||
NM_020313.3 | NP_064709.2 | 9 | 1776 | |||
XM_005256063.4 | XP_005256120.1 | 8 | 1512 | |||
GenBank mRNA | AF116609.1 | - | 7 | 1387 | ||
AF248964.1 | - | 9 | 1680 | |||
AK074263.1 | - | 9 | 1617 | |||
AL136613.1 | - | 9 | 1598 | |||
BC002568.2 | - | 9 | 1618 | 3139226 | ||
BC024196.2 | - | 9 | 1657 | 4816594 | ||
BC067303.1 | - | 9 | 1776 | 30389410 | ||
BC071740.1 | - | 9 | 1651 | 5563094 |