Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020748.2 | NP_065799.1 | 8 | 1151 | ||
NR_026641.1 | 8 | 1432 | ||||
XM_006721987.3 | XP_006722050.1 | 8 | 1158 | |||
XM_011525058.2 | XP_011523360.1 | 8 | 1438 | |||
GenBank mRNA | AB033113.1 | - | 8 | 1379 | ||
AK299032.1 | - | 8 | 1407 | |||
AK308307.1 | - | 8 | 1116 | |||
AL136800.1 | - | 8 | 1151 | |||
CR533582.1 | - | 7 | 1075 |