Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020774.3 | NP_065825.1 | 4 | 826 | ||
XM_017025874.1 | XP_016881363.1 | 4 | 1362 | |||
XM_017025875.1 | XP_016881364.1 | 4 | 1362 | |||
GenBank mRNA | AK091266.1 | - | 3 | 256 | ||
AY147849.1 | - | 4 | 844 | |||
AY149908.1 | - | 4 | 571 | |||
BC110581.1 | - | 4 | 571 | 40008394 | ||
BC110582.1 | - | 4 | 571 | 40008395 |