Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130136.1 | NP_001123608.1 | 4 | 2505 | ||
NM_001130137.1 | NP_001123609.1 | 4 | 2241 | |||
NM_001130138.1 | NP_001123610.1 | 4 | 1949 | |||
NM_020851.2 | NP_065902.1 | 3 | 2074 | |||
XM_011521840.2 | XP_011520142.1 | 4 | 2344 | |||
XM_011521841.1 | XP_011520143.1 | 4 | 2027 | |||
GenBank mRNA | AB040898.2 | - | 4 | 2505 | ||
AK092297.1 | - | 3 | 1900 | |||
AK092302.1 | - | 4 | 2521 | |||
AK096602.1 | - | 2 | 1852 | |||
AK290467.1 | - | 3 | 1943 | |||
AY358316.1 | - | 3 | 1872 | |||
BC028042.1 | - | 4 | 2237 | |||
BC114551.1 | - | 3 | 588 | |||
BC152429.1 | - | 4 | 2505 | 8862545 |