Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130136.1 | NP_001123608.1 | 4 | 3249 | ||
NM_001130137.1 | NP_001123609.1 | 4 | 2985 | |||
NM_001130138.1 | NP_001123610.1 | 4 | 2693 | |||
NM_020851.2 | NP_065902.1 | 3 | 2818 | |||
XM_011521840.2 | XP_011520142.1 | 4 | 3088 | |||
XM_011521841.1 | XP_011520143.1 | 4 | 2771 | |||
GenBank mRNA | AB040898.2 | - | 4 | 3249 | ||
AK092297.1 | - | 3 | 2644 | |||
AK092302.1 | - | 4 | 3265 | |||
AK096602.1 | - | 2 | 2596 | |||
AY358316.1 | - | 3 | 2616 | |||
BC114551.1 | - | 3 | 1332 | |||
BC152429.1 | - | 4 | 3249 | 8862545 |