Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020947.3 | NP_065998.3 | 8 | 1792 | ||
XM_005256074.3 | XP_005256131.1 | 8 | 1862 | |||
XM_005256075.2 | XP_005256132.1 | 9 | 1990 | |||
XM_006721240.2 | XP_006721303.1 | 9 | 1945 | |||
XM_011523248.2 | XP_011521550.1 | 10 | 2089 | |||
XM_017023510.1 | XP_016878999.1 | 10 | 2109 | |||
XM_017023511.1 | XP_016879000.1 | 8 | 1864 | |||
XM_017023512.1 | XP_016879001.1 | 9 | 2032 | |||
GenBank mRNA | AB046829.2 | - | 1737 | |||
AK297241.1 | - | 4 | 926 | |||
AK310887.1 | - | 8 | 1868 | |||
AL137316.1 | - | 3 | 661 | |||
BC060844.1 | - | 8 | 1779 | 30342219 | ||
BG748167.1 | - | 2 | 488 |