Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 12 | 2538 | ||
NM_020954.3 | NP_066005.2 | 12 | 2538 | |||
XM_005257545.4 | XP_005257602.2 | 13 | 2726 | |||
XM_005257546.4 | XP_005257603.2 | 13 | 2630 | |||
XM_006721995.3 | XP_006722058.1 | 13 | 3013 | |||
XM_011525084.2 | XP_011523386.1 | 13 | 2726 | |||
XM_011525086.2 | XP_011523388.1 | 13 | 2726 | |||
XM_011525087.2 | XP_011523389.1 | 13 | 2726 | |||
XM_017024905.1 | XP_016880394.1 | 8 | 1593 | |||
GenBank mRNA | AB046838.1 | - | 9 | 1983 | ||
AB537889.1 | - | 12 | 2531 | |||
AK023113.1 | - | 1 | 64 | |||
AL832920.1 | - | 12 | 2525 | |||
BC036891.1 | - | 12 | 2554 | 5501834 | ||
BC040341.1 | - | 12 | 2529 | 3873001 |