Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGDCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020962.2 | NP_066013.1 | 20 | 5706 | ||
XM_011521845.2 | XP_011520147.1 | 20 | 5886 | |||
XM_011521846.2 | XP_011520148.1 | 20 | 5513 | |||
XM_011521847.1 | XP_011520149.1 | 20 | 5573 | |||
XM_017022448.1 | XP_016877937.1 | 20 | 5891 | |||
GenBank mRNA | AB046848.1 | - | 14 | 4770 | ||
AB052622.1 | - | 20 | 5706 | |||
AK124045.1 | - | 3 | 2628 | |||
BC040648.1 | - | 1 | 1767 |